5-(3,4-Dimethoxyphenyl)nicotinic acid

Lieferant: Apollo Scientific

OR470376-5G
APOSOR470376-5GEA 1940 EUR
APOSOR470376-5G
5-(3,4-Dimethoxyphenyl)nicotinic acid
5-(3,4-Dimethoxyphenyl)nicotinic acid
Formel: C₁₄H₁₃NO₄
Molecular Weight: 259.257 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06410296
CAS-Nummer: 887973-42-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR