N-(4-Bromobenzyl)-2,2-dimethoxyethylamine

Lieferant: Apollo Scientific

OR470380-5G
APOSOR470380-5GEA 601 EUR
APOSOR470380-5G
N-(4-Bromobenzyl)-2,2-dimethoxyethylamine
N-(4-Bromobenzyl)-2,2-dimethoxyethylamine
Formel: C₁₁H₁₆BrNO₂
Molecular Weight: 274,154 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12148381
CAS-Nummer: 1036378-89-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR