2-[2-(Vinyloxy)ethoxy]isoindoline-1,3-dione

Lieferant: Apollo Scientific

Synonyme: 2-(2-(Vinyloxy)ethoxy)isoindoline-1,3-dione

OR470381-5G
APOSOR470381-5GEA 916 EUR
APOSOR470381-5G
2-[2-(Vinyloxy)ethoxy]isoindoline-1,3-dione
2-[2-(Vinyloxy)ethoxy]isoindoline-1,3-dione
Formel: C₁₂H₁₁NO₄
Molecular Weight: 233,22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD15142583
CAS-Nummer: 391212-30-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR