N-(2-Chlorophenyl)cinnamamide

Lieferant: Apollo Scientific

OR470382-5G
APOSOR470382-5GEA 448 EUR
APOSOR470382-5G
N-(2-Chlorophenyl)cinnamamide
N-(2-Chlorophenyl)cinnamamide
Formel: C₁₅H₁₂ClNO
Molecular Weight: 257,715 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00157758
CAS-Nummer: 73108-79-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR