4-Acetamido-3,5-dimethylphenol

Lieferant: Apollo Scientific

OR470383-5G
APOSOR470383-5GEA 713 EUR
APOSOR470383-5G
4-Acetamido-3,5-dimethylphenol
4-Acetamido-3,5-dimethylphenol
Formel: C₁₀H₁₃NO₂
Molecular Weight: 179,216 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00748645
CAS-Nummer: 6337-56-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR