6-Cbz-2,2-dimethylhexahydropyrrolo[3,4-d]-1,3-oxazine
Lieferant: Apollo Scientific
OR470384-1G
APOSOR470384-1GEA
896
EUR
APOSOR470384-1G
6-Cbz-2,2-dimethylhexahydropyrrolo[3,4-d]-1,3-oxazine
6-Cbz-2,2-dimethylhexahydropyrrolo[3,4-d]-1,3-oxazine
Formel:
C₁₆H₂₂N₂O₃ Molecular Weight: 290.357 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD20134214 CAS-Nummer: 1346597-57-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...