3-Amino-3-(3-tert-butoxyphenyl)propanoic acid

Lieferant: Apollo Scientific

OR470386-5G
APOSOR470386-5GEA 1540 EUR
APOSOR470386-5G
3-Amino-3-(3-tert-butoxyphenyl)propanoic acid
3-Amino-3-(3-tert-butoxyphenyl)propanoic acid
Formel: C₁₃H₁₉NO₃
Molecular Weight: 237,295 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02683181
CAS-Nummer: 372144-15-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR