5-(3-Methylphenyl)thiophene-2-carbaldehyde

Lieferant: Apollo Scientific

OR470389-5G
APOSOR470389-5GEA 997 EUR
APOSOR470389-5G
5-(3-Methylphenyl)thiophene-2-carbaldehyde
5-(3-Methylphenyl)thiophene-2-carbaldehyde
Formel: C₁₂H₁₀OS
Molecular Weight: 202,272 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06410188
CAS-Nummer: 38401-69-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR