3-(Boc-amino)-3-(4-chlorophenyl)-N-hydroxypropanamide

Lieferant: Apollo Scientific

OR470392-5G
APOSOR470392-5GEA 1620 EUR
APOSOR470392-5G
3-(Boc-amino)-3-(4-chlorophenyl)-N-hydroxypropanamide
3-(Boc-amino)-3-(4-chlorophenyl)-N-hydroxypropanamide
Formel: C₁₄H₁₉ClN₂O₄
Molecular Weight: 314.765 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04115559
CAS-Nummer: 898404-71-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR