2-(4-Biphenylyl)-2-(Boc-amino)acetic acid

Lieferant: Apollo Scientific

OR470393-5G
APOSOR470393-5GEA 1620 EUR
APOSOR470393-5G
2-(4-Biphenylyl)-2-(Boc-amino)acetic acid
2-(4-Biphenylyl)-2-(Boc-amino)acetic acid
Formel: C₁₉H₂₁NO₄
Molecular Weight: 327,374 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02683167
CAS-Nummer: 369403-44-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR