1-(3,4-Dimethoxyphenyl)-4-oxocyclohexanecarboxylic acid

Lieferant: Apollo Scientific

OR470394-5G
APOSOR470394-5GEA 1010 EUR
APOSOR470394-5G
1-(3,4-Dimethoxyphenyl)-4-oxocyclohexanecarboxylic acid
1-(3,4-Dimethoxyphenyl)-4-oxocyclohexanecarboxylic acid
Formel: C₁₅H₁₈O₅
Molecular Weight: 278,3 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06410575
CAS-Nummer: 887978-64-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR