Fmoc-2-amino-5-chlorobenzoic acid

Lieferant: Apollo Scientific

OR470395-5G
APOSOR470395-5GEA 499 EUR
APOSOR470395-5G
Fmoc-2-amino-5-chlorobenzoic acid
Fmoc-2-amino-5-chlorobenzoic acid
Formel: C₂₂H₁₆ClNO₄
Molecular Weight: 393,83 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02682159
CAS-Nummer: 332121-90-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR