Ethyl 3-(Bicyclo[2.2.1]hept-5-en-2-yl)-3-oxopropanoate

Lieferant: Apollo Scientific

OR470397-5G
APOSOR470397-5GEA 1120 EUR
APOSOR470397-5G
Ethyl 3-(Bicyclo[2.2.1]hept-5-en-2-yl)-3-oxopropanoate
Ethyl 3-(Bicyclo[2.2.1]hept-5-en-2-yl)-3-oxopropanoate
Formel: C₁₂H₁₆O₃
Molecular Weight: 208,254 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10465036
CAS-Nummer: 75275-66-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR