1-Boc-4-[(benzylamino)methyl]piperidine

Lieferant: Apollo Scientific

Synonyme: tert-Butyl 4-((benzylamino)methyl)piperidine-1-carboxylate

OR470398-5G
APOSOR470398-5GEA 1120 EUR
APOSOR470398-5G
1-Boc-4-[(benzylamino)methyl]piperidine
1-Boc-4-[(benzylamino)methyl]piperidine
Formel: C₁₈H₂₈N₂O₂
Molecular Weight: 304,427 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06410625
CAS-Nummer: 195314-72-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR