2-Amino-2-(6-phenyl-3-pyridyl)acetic acid hydrochloride

Lieferant: Apollo Scientific

OR470399-5G
APOSOR470399-5GEA 1840 EUR
APOSOR470399-5G
2-Amino-2-(6-phenyl-3-pyridyl)acetic acid hydrochloride
2-Amino-2-(6-phenyl-3-pyridyl)acetic acid hydrochloride
Formel: C₁₃H₁₂N₂O₂·ClH
Molecular Weight: 264,708 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07371470
CAS-Nummer: 1134915-37-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR