(E)-3-(3-tert-Butoxyphenyl)acrylic acid
Lieferant: Apollo Scientific
OR470400-5G
APOSOR470400-5GEA
1330
EUR
APOSOR470400-5G
(E)-3-(3-tert-Butoxyphenyl)acrylic acid
(E)-3-(3-tert-Butoxyphenyl)acrylic acid
Formel:
C₁₃H₁₆O₃ Molecular Weight: 220,264 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03412398 CAS-Nummer: 898405-11-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...