O-(3-Pyridyl)-L-tyrosine methyl ester

Lieferant: Apollo Scientific

OR470401-5G
APOSOR470401-5GEA 1330 EUR
APOSOR470401-5G
O-(3-Pyridyl)-L-tyrosine methyl ester
O-(3-Pyridyl)-L-tyrosine methyl ester
Formel: C₁₅H₁₆N₂O₃
Molecular Weight: 272.299 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD20922889
CAS-Nummer: 1137013-14-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR