Ethyl 3-Oxo-3-(1-Cbz-3-piperidyl)propanoate

Lieferant: Apollo Scientific

OR470404-5G
APOSOR470404-5GEA 1330 EUR
APOSOR470404-5G
Ethyl 3-Oxo-3-(1-Cbz-3-piperidyl)propanoate
Ethyl 3-Oxo-3-(1-Cbz-3-piperidyl)propanoate
Formel: C₁₈H₂₃NO₅
Molecular Weight: 333,379 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04115572
CAS-Nummer: 672323-13-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR