4-Aminotetrahydro-2H-thiopyran-4-carboxylic acid
Lieferant: Apollo Scientific
OR470405-5G
APOSOR470405-5GEA
1280
EUR
APOSOR470405-5G
4-Aminotetrahydro-2H-thiopyran-4-carboxylic acid
4-Aminotetrahydro-2H-thiopyran-4-carboxylic acid
Formel:
C₆H₁₁NO₂S Molecular Weight: 161,22 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06656113 CAS-Nummer: 39124-16-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...