2-Amino-2-(3,4,5-trimethoxyphenyl)acetic acid hydrochloride
Lieferant: Apollo Scientific
OR470406-5G
APOSOR470406-5GEA
916
EUR
APOSOR470406-5G
2-Amino-2-(3,4,5-trimethoxyphenyl)acetic acid hydrochloride
2-Amino-2-(3,4,5-trimethoxyphenyl)acetic acid hydrochloride
Formel:
C₁₁H₁₅NO₅·ClH Lagertemperatur: Raumtemperatur |
MDL:
MFCD06410544 CAS-Nummer: 86053-95-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...