5-(2-Methoxyphenyl)nicotinic acid

Lieferant: Apollo Scientific

OR470407-5G
APOSOR470407-5GEA 1540 EUR
APOSOR470407-5G
5-(2-Methoxyphenyl)nicotinic acid
5-(2-Methoxyphenyl)nicotinic acid
Formel: C₁₃H₁₁NO₃
Molecular Weight: 229.231 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06410272
CAS-Nummer: 893732-61-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR