3-tert-Butoxybenzaldehyde

Lieferant: Apollo Scientific

OR470408-5G
APOSOR470408-5GEA 804 EUR
APOSOR470408-5G
3-tert-Butoxybenzaldehyde
3-tert-Butoxybenzaldehyde
Formel: C₁₁H₁₄O₂
Molecular Weight: 178,228 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02258842
CAS-Nummer: 64859-36-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR