1-(4-Methoxyphenyl)-4-oxocyclohexanecarboxylic acid

Lieferant: Apollo Scientific

OR470409-5G
APOSOR470409-5GEA 1010 EUR
APOSOR470409-5G
1-(4-Methoxyphenyl)-4-oxocyclohexanecarboxylic acid
1-(4-Methoxyphenyl)-4-oxocyclohexanecarboxylic acid
Formel: C₁₄H₁₆O₄
Molecular Weight: 248,274 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06410573
CAS-Nummer: 887978-56-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR