1-(3,4-Dichlorophenyl)-4-oxocyclohexanecarboxylic acid

Lieferant: Apollo Scientific

OR470410-5G
APOSOR470410-5GEA 1010 EUR
APOSOR470410-5G
1-(3,4-Dichlorophenyl)-4-oxocyclohexanecarboxylic acid
1-(3,4-Dichlorophenyl)-4-oxocyclohexanecarboxylic acid
Formel: C₁₃H₁₂Cl₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD06208895
CAS-Nummer: 773101-05-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR