Tri-tert-butylmethanol

Lieferant: Apollo Scientific

Synonyme: 3-(Tert-butyl)-2,2,4,4-tetramethylpentan-3-ol

OR470411-250MG
APOSOR470411-250MGEA 235 EUR
APOSOR470411-250MG
Tri-tert-butylmethanol
Tri-tert-butylmethanol
Formel: C₁₃H₂₈O
Molecular Weight: 200.361 g/mol
Schmelzpunkt: 94…95 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00204381
CAS-Nummer: 41902-42-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR