Methyl 4-(3-Bromophenyl)-2,4-dioxobutanoate

Lieferant: Apollo Scientific

OR470412-1G
APOSOR470412-1GEA 601 EUR
APOSOR470412-1G
Methyl 4-(3-Bromophenyl)-2,4-dioxobutanoate
Methyl 4-(3-Bromophenyl)-2,4-dioxobutanoate
Formel: C₁₁H₉BrO₄
Molecular Weight: 285,091 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04969738
CAS-Nummer: 93618-22-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR