2-[4-(Boc-amino)-3-pyridyl]ethanol

Lieferant: Apollo Scientific

OR470413-1G
APOSOR470413-1GEA 591 EUR
APOSOR470413-1G
2-[4-(Boc-amino)-3-pyridyl]ethanol
2-[4-(Boc-amino)-3-pyridyl]ethanol
Formel: C₁₂H₁₈N₂O₃
Molecular Weight: 238,283 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20491404
CAS-Nummer: 219834-80-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR