1-Boc-2-(2-methoxy-2-oxoethylcarbamoyl)pyrrolidine

Lieferant: Apollo Scientific

OR470415-5G
APOSOR470415-5GEA 601 EUR
APOSOR470415-5G
1-Boc-2-(2-methoxy-2-oxoethylcarbamoyl)pyrrolidine
1-Boc-2-(2-methoxy-2-oxoethylcarbamoyl)pyrrolidine
Formel: C₁₃H₂₂N₂O₅
Molecular Weight: 286,324 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01046139
CAS-Nummer: 1097194-13-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR