Ethyl 3-(2-Bromo-6-nitrophenyl)-2-oxopropanoate

Lieferant: Apollo Scientific

OR470416-5G
APOSOR470416-5GEA 397 EUR
APOSOR470416-5G
Ethyl 3-(2-Bromo-6-nitrophenyl)-2-oxopropanoate
Ethyl 3-(2-Bromo-6-nitrophenyl)-2-oxopropanoate
Formel: C₁₁H₁₀BrNO₅
Molecular Weight: 316,105 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20491407
CAS-Nummer: 608510-29-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR