N-Benzyl-N-methyl-1H-benzotriazole-1-methanamine

Lieferant: Apollo Scientific

OR470418-5G
APOSOR470418-5GEA 397 EUR
APOSOR470418-5G
N-Benzyl-N-methyl-1H-benzotriazole-1-methanamine
N-Benzyl-N-methyl-1H-benzotriazole-1-methanamine
Formel: C₁₅H₁₆N₄
Molecular Weight: 252,314 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00964110
CAS-Nummer: 57684-27-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR