5-[(3-Methoxyphenylamino)methylene]-2,2-dimethyl-1,3-dioxane-4,6-dione

Lieferant: Apollo Scientific

OR470421-25G
APOSOR470421-25GEA 601 EUR
APOSOR470421-25G
5-[(3-Methoxyphenylamino)methylene]-2,2-dimethyl-1,3-dioxane-4,6-dione
5-[(3-Methoxyphenylamino)methylene]-2,2-dimethyl-1,3-dioxane-4,6-dione
Formel: C₁₄H₁₅NO₅
Molecular Weight: 277,273 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02936591
CAS-Nummer: 213699-52-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR