5-(4-Methoxyphenyl)imidazolidine-2,4-dione

Lieferant: Apollo Scientific

OR470423-5G
APOSOR470423-5GEA 397 EUR
APOSOR470423-5G
5-(4-Methoxyphenyl)imidazolidine-2,4-dione
5-(4-Methoxyphenyl)imidazolidine-2,4-dione
Formel: C₁₀H₁₀N₂O₃
Molecular Weight: 206,198 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00219347
CAS-Nummer: 6617-78-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR