6-Pivalamidopyridine-3-boronic acid pinacol ester

Lieferant: Apollo Scientific

OR470426-1G
APOSOR470426-1GEA 804 EUR
APOSOR470426-1G
6-Pivalamidopyridine-3-boronic acid pinacol ester
6-Pivalamidopyridine-3-boronic acid pinacol ester
Formel: C₁₆H₂₅BN₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD13191341
CAS-Nummer: 1092119-20-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR