Methyl 4-(Acetoxymethyl)benzoate

Lieferant: Apollo Scientific

OR470427-5G
APOSOR470427-5GEA 601 EUR
APOSOR470427-5G
Methyl 4-(Acetoxymethyl)benzoate
Methyl 4-(Acetoxymethyl)benzoate
Formel: C₁₁H₁₂O₄
Molecular Weight: 208,211 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21332892
CAS-Nummer: 27548-25-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR