2-(Cyclohexyloxy)acetamide

Lieferant: Apollo Scientific

OR470431-5G
APOSOR470431-5GEA 672 EUR
APOSOR470431-5G
2-(Cyclohexyloxy)acetamide
2-(Cyclohexyloxy)acetamide
Formel: C₈H₁₅NO₂
Molecular Weight: 157,21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16042596
CAS-Nummer: 98552-43-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR