N-Methyl-N-(methylsulphonyl)glycine ethyl ester
Lieferant: Apollo Scientific
OR470432-5G
APOSOR470432-5GEA
601
EUR
APOSOR470432-5G
N-Methyl-N-(methylsulphonyl)glycine ethyl ester
N-Methyl-N-(methylsulphonyl)glycine ethyl ester
Formel:
C₆H₁₃NO₄S Molecular Weight: 195,237 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD16042210 CAS-Nummer: 58742-72-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...