Ethyl 4-(Methylsulphonyl)-2-morpholinobenzoate
Lieferant: Apollo Scientific
OR470433-5G
APOSOR470433-5GEA
997
EUR
APOSOR470433-5G
Ethyl 4-(Methylsulphonyl)-2-morpholinobenzoate
Ethyl 4-(Methylsulphonyl)-2-morpholinobenzoate
Formel:
C₁₄H₁₉NO₅S Lagertemperatur: Raumtemperatur |
MDL:
MFCD21332900 CAS-Nummer: 1354940-64-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...