3-Hydroxy-4-methoxy-5-nitrobenzaldehyde

Lieferant: Apollo Scientific

OR470435-1G
APOSOR470435-1GEA 804 EUR
APOSOR470435-1G
3-Hydroxy-4-methoxy-5-nitrobenzaldehyde
3-Hydroxy-4-methoxy-5-nitrobenzaldehyde
Formel: C₈H₇NO₅
Molecular Weight: 197,145 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02380161
CAS-Nummer: 80547-69-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR