2-(2-Cyanoethylamino)-2-methylpropanoic acid 97%

Lieferant: Apollo Scientific
Achtung

OR470437-1G
APOSOR470437-1GEA 139 EUR
APOSOR470437-1G
2-(2-Cyanoethylamino)-2-methylpropanoic acid 97%
2-(2-Cyanoethylamino)-2-methylpropanoic acid
Formel: C₇H₁₂N₂O₂
Molecular Weight: 156,182 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD12756521
CAS-Nummer: 106556-63-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR