2-Amino-1,3,5-triazino[1,2-a]benzimidazol-4(3H)-one

Lieferant: Apollo Scientific

OR470439-5G
APOSOR470439-5GEA 804 EUR
APOSOR470439-5G
2-Amino-1,3,5-triazino[1,2-a]benzimidazol-4(3H)-one
2-Amino-1,3,5-triazino[1,2-a]benzimidazol-4(3H)-one
Formel: C₉H₇N₅O
Lagertemperatur: Raumtemperatur
MDL: MFCD02252785
CAS-Nummer: 52066-00-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR