3,4-Dihydro-1,3,5-triazino[1,2-a]benzimidazol-2-amine

Lieferant: Apollo Scientific

OR470441-5G
APOSOR470441-5GEA 804 EUR
APOSOR470441-5G
3,4-Dihydro-1,3,5-triazino[1,2-a]benzimidazol-2-amine
3,4-Dihydro-1,3,5-triazino[1,2-a]benzimidazol-2-amine
Formel: C₉H₉N₅
Molecular Weight: 187,201 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02323005
CAS-Nummer: 78650-01-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR