3,4-Dihydro-4-phenyl-1,3,5-triazino[1,2-a]benzimidazol-2-amine
Lieferant: Apollo Scientific
OR470442-5G
APOSOR470442-5GEA
804
EUR
APOSOR470442-5G
3,4-Dihydro-4-phenyl-1,3,5-triazino[1,2-a]benzimidazol-2-amine
3,4-Dihydro-4-phenyl-1,3,5-triazino[1,2-a]benzimidazol-2-amine
Formel:
C₁₅H₁₃N₅ Molecular Weight: 263,297 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00188404 CAS-Nummer: 26958-67-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...