3,4-Dihydro-4,4-dimethyl-1,3,5-triazino[1,2-a]benzimidazol-2-amine

Lieferant: Apollo Scientific
Achtung

OR470443-5G
APOSOR470443-5GEA 804 EUR
APOSOR470443-5G
3,4-Dihydro-4,4-dimethyl-1,3,5-triazino[1,2-a]benzimidazol-2-amine
3,4-Dihydro-4,4-dimethyl-1,3,5-triazino[1,2-a]benzimidazol-2-amine
Formel: C₁₁H₁₃N₅
Molecular Weight: 215,254 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02323006
CAS-Nummer: 325738-83-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR