2-Amino-1,3,5-triazino[1,2-a]benzimidazole-4(3H)-thione
Lieferant: Apollo Scientific
OR470444-5G
APOSOR470444-5GEA
1010
EUR
APOSOR470444-5G
2-Amino-1,3,5-triazino[1,2-a]benzimidazole-4(3H)-thione
2-Amino-1,3,5-triazino[1,2-a]benzimidazole-4(3H)-thione
Formel:
C₉H₇N₅S Molecular Weight: 217,25 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02252796 CAS-Nummer: 78650-21-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...