Ethyl 2-(2-Benzimidazolylamino)-6-oxo-1,6-dihydropyrimidine-5-carboxylate

Lieferant: Apollo Scientific

OR470445-5G
APOSOR470445-5GEA 804 EUR
APOSOR470445-5G
Ethyl 2-(2-Benzimidazolylamino)-6-oxo-1,6-dihydropyrimidine-5-carboxylate
Ethyl 2-(2-Benzimidazolylamino)-6-oxo-1,6-dihydropyrimidine-5-carboxylate
Formel: C₁₄H₁₃N₅O₃
Molecular Weight: 299,285 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22205839
CAS-Nummer: 195251-35-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR