2-(2-Benzimidazolylamino)-4-chloro-6-methylpyrimidine

Lieferant: Apollo Scientific

OR470447-5G
APOSOR470447-5GEA 1410 EUR
APOSOR470447-5G
2-(2-Benzimidazolylamino)-4-chloro-6-methylpyrimidine
2-(2-Benzimidazolylamino)-4-chloro-6-methylpyrimidine
Formel: C₁₂H₁₀ClN₅
Molecular Weight: 259,694 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22205840
CAS-Nummer: 42389-34-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR