Ethyl 2-(2-Benzoxazolylamino)-6-oxo-1,6-dihydropyrimidine-5-carboxylate
Lieferant: Apollo Scientific
OR470448-5G
APOSOR470448-5GEA
804
EUR
APOSOR470448-5G
Ethyl 2-(2-Benzoxazolylamino)-6-oxo-1,6-dihydropyrimidine-5-carboxylate
Ethyl 2-(2-Benzoxazolylamino)-6-oxo-1,6-dihydropyrimidine-5-carboxylate
Formel:
C₁₄H₁₂N₄O₄ Lagertemperatur: Raumtemperatur |
MDL:
MFCD22205841 CAS-Nummer: 670220-69-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...