2-(1,3-Benzoxazol-2-ylamino)-6-methylpyrimidin-4-ol

Lieferant: Apollo Scientific

Synonyme: 2-(2-Benzoxazolylamino)-6-methylpyrimidin-4(3H)-one

OR470449-5G
APOSOR470449-5GEA 1010 EUR
APOSOR470449-5G
2-(1,3-Benzoxazol-2-ylamino)-6-methylpyrimidin-4-ol
2-(1,3-Benzoxazol-2-ylamino)-6-methylpyrimidin-4-ol
Formel: C₁₂H₁₀N₄O₂
Molecular Weight: 242,24 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01948713
CAS-Nummer: 86328-33-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR