Ethyl 2-Oxo-5-phenyl-2,3-dihydro-1H-imidazole-4-carboxylate
Lieferant: Apollo Scientific
OR470450-1G
APOSOR470450-1GEA
713
EUR
APOSOR470450-1G
Ethyl 2-Oxo-5-phenyl-2,3-dihydro-1H-imidazole-4-carboxylate
Ethyl 2-Oxo-5-phenyl-2,3-dihydro-1H-imidazole-4-carboxylate
Formel:
C₁₂H₁₂N₂O₃ Molecular Weight: 232,235 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22574973 CAS-Nummer: 92809-78-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...