(R)-1-Boc-2-propylpiperazine hydrochloride
Lieferant: Apollo Scientific
Synonyme:
(R)-tert-Butyl 2-propylpiperazine-1-carboxylate hydrochloride
OR470453-1G
APOSOR470453-1GEA
277
EUR
APOSOR470453-1G
(R)-1-Boc-2-propylpiperazine hydrochloride
(R)-1-Boc-2-propylpiperazine hydrochloride
Formel:
C₁₂H₂₄N₂O₂·ClH Molecular Weight: 264,792 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11101218 CAS-Nummer: 1217449-00-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...